Assay Details
| Gene Symbol: Akt1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510880.2 | XP_003510928.2 | 2 - 3 | 966 | 57 | |
| XM_007651468.1 | XP_007649658.1 | 3 - 4 | 1177 | 57 | ||
| XM_007651469.1 | XP_007649659.1 | 4 - 5 | 812 | 57 | ||
| GenBank mRNA | JI862337.1 | - | 1 - 2 | 93 | 57 | |
More Information
Set Membership: |
|

Akt1s1
Genomic Map