Assay Details
Gene Symbol: LOC100759042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642550.1 | XP_007640740.1 | 4 - 5 | 746 | 111 | |
XM_007642551.1 | XP_007640741.1 | 5 - 6 | 808 | 111 | ||
XM_007642552.1 | XP_007640742.1 | 5 - 6 | 714 | 111 | ||
XM_007642553.1 | XP_007640743.1 | 4 - 5 | 387 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
