Assay Details
| Gene Symbol: LOC100759042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642550.1 | XP_007640740.1 | 5 - 6 | 1887 | 88 | |
| XM_007642551.1 | XP_007640741.1 | 6 - 7 | 1949 | 88 | ||
| XM_007642552.1 | XP_007640742.1 | 6 - 7 | 1855 | 88 | ||
| XM_007642553.1 | XP_007640743.1 | 5 - 6 | 1528 | 88 | ||
More Information
Set Membership: |
|

LOC100759042
Genomic Map