Assay Details
| Gene Symbol: LOC100767606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652143.1 | XP_007650333.1 | 19 - 20 | 2685 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100767606| Gene Symbol: LOC100767606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652143.1 | XP_007650333.1 | 19 - 20 | 2685 | 85 | |
Set Membership: |
|