Assay Details
| Gene Symbol: Il10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501219.2 | XP_003501267.1 | 1 - 2 | 980 | 69 | |
| XM_007641911.1 | XP_007640101.1 | 1 - 2 | 1006 | 69 | ||
More Information
Set Membership: |
|

Il10
Genomic Map