Assay Details
Gene Symbol: Lect1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511312.1 | XP_003511360.1 | 5 - 6 | 617 | 89 | |
XM_007651839.1 | XP_007650029.1 | 5 - 6 | 617 | 89 | ||
XM_007651840.1 | XP_007650030.1 | 5 - 6 | 617 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
