Assay Details
| Gene Symbol: Krt72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512317.1 | XP_003512365.1 | 2 - 3 | 484 | 65 | |
| XM_003512318.1 | XP_003512366.1 | 2 - 3 | 484 | 65 | ||
More Information
Set Membership: |
|

Krt72
Genomic Map