Assay Details
| Gene Symbol: Krt72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512317.1 | XP_003512365.1 | 6 - 7 | 833 | 84 | |
| XM_003512318.1 | XP_003512366.1 | 6 - 7 | 833 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt72| Gene Symbol: Krt72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512317.1 | XP_003512365.1 | 6 - 7 | 833 | 84 | |
| XM_003512318.1 | XP_003512366.1 | 6 - 7 | 833 | 84 | ||