Assay Details
| Gene Symbol: Loxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643026.1 | XP_007641216.1 | 3 - 4 | 755 | 72 | |
| GenBank mRNA | JI893127.1 | - | 2 - 3 | 483 | 72 | |
More Information
Set Membership: |
|

Loxl1
Genomic Map