Assay Details
| Gene Symbol: Spag4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641854.1 | XP_007640044.1 | 9 - 10 | 913 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spag4| Gene Symbol: Spag4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641854.1 | XP_007640044.1 | 9 - 10 | 913 | 59 | |
Set Membership: |
|