Assay Details
| Gene Symbol: Zfand4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647944.1 | XP_007646134.1 | 7 - 8 | 1081 | 86 | |
| XM_007647945.1 | XP_007646135.1 | 6 - 7 | 974 | 86 | ||
| XM_007647946.1 | XP_007646136.1 | 7 - 8 | 1081 | 86 | ||
| XM_007647948.1 | XP_007646138.1 | 7 - 8 | 1081 | 86 | ||
| XM_007647949.1 | XP_007646139.1 | 4 - 5 | 460 | 86 | ||
| XM_007647950.1 | XP_007646140.1 | 3 - 4 | 249 | 86 | ||
More Information
Set Membership: |
|

Zfand4
Genomic Map