Assay Details
| Gene Symbol: Ncr3lg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649817.1 | XP_007648007.1 | 1 - 2 | 348 | 113 | |
| XM_007649818.1 | XP_007648008.1 | 1 - 2 | 348 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_007649817.1, XM_007649818.1 transcripts.
Set Membership: |
|

Ncr3lg1
Genomic Map