Assay Details
| Gene Symbol: Ncr3lg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649817.1 | XP_007648007.1 | 3 - 4 | 1005 | 85 | |
| XM_007649818.1 | XP_007648008.1 | 3 - 4 | 1005 | 85 | ||
More Information
Set Membership: |
|

Ncr3lg1
Genomic Map