Assay Details
| Gene Symbol: Rybp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495219.2 | XP_003495267.2 | 1 - 2 | 244 | 60 | |
| GenBank mRNA | JI864333.1 | - | 1 - 2 | 244 | 60 | |
More Information
Set Membership: |
|

Rybp
Genomic Map