Assay Details
| Gene Symbol: Dynll2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498196.2 | XP_003498244.1 | 2 - 3 | 159 | 95 | |
| XM_007624215.1 | XP_007622405.1 | 2 - 3 | 408 | 95 | ||
| GenBank mRNA | JI868689.1 | - | 2 - 3 | 444 | 95 | |
| JP038068.1 | - | 2 - 3 | 444 | 95 | ||
More Information
Set Membership: |
|

Dynll2
Genomic Map