Assay Details
| Gene Symbol: Art4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499215.1 | XP_003499263.1 | 2 - 3 | 795 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Art4| Gene Symbol: Art4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499215.1 | XP_003499263.1 | 2 - 3 | 795 | 88 | |
Set Membership: |
|