Assay Details
| Gene Symbol: Nudt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500661.1 | XP_003500709.1 | 1 - 2 | 38 | 56 | |
| XM_003500662.1 | XP_003500710.1 | 1 - 2 | 38 | 56 | ||
More Information
Set Membership: |
|

Nudt7
Genomic Map