Assay Details
| Gene Symbol: Rpp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502729.2 | XP_003502777.2 | 4 - 5 | 473 | 68 | |
| XM_007643345.1 | XP_007641535.1 | 4 - 5 | 376 | 68 | ||
| XM_007643346.1 | XP_007641536.1 | 4 - 5 | 276 | 68 | ||
| GenBank mRNA | JP054914.1 | - | 3 - 4 | 268 | 68 | |
More Information
Set Membership: |
|

Rpp14
Genomic Map