Assay Details
| Gene Symbol: Tubg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504660.2 | XP_003504708.1 | 10 - 11 | 1234 | 80 | |
| GenBank mRNA | JI875183.1 | - | 3 - 4 | 469 | 80 | |
| JP063301.1 | - | 10 - 11 | 1250 | 80 | ||
More Information
Set Membership: |
|

Tubg1
Genomic Map