Assay Details
| Gene Symbol: Nif3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507030.2 | XP_003507078.1 | 6 - 7 | 1028 | 63 | |
| XM_007647580.1 | XP_007645770.1 | 5 - 6 | 1255 | 63 | ||
| XM_007647581.1 | XP_007645771.1 | 4 - 5 | 1118 | 63 | ||
| GenBank mRNA | JI890539.1 | - | 6 - 7 | 995 | 63 | |
More Information
Set Membership: |
|

Nif3l1
Genomic Map