Assay Details
| Gene Symbol: Uxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507943.2 | XP_003507991.1 | 6 - 7 | 484 | 69 | |
| XM_007648524.1 | XP_007646714.1 | 6 - 7 | 442 | 69 | ||
| GenBank mRNA | JI890343.1 | - | 4 - 5 | 317 | 69 | |
| JP064423.1 | - | 6 - 7 | 460 | 69 | ||
More Information
Set Membership: |
|

Uxt
Genomic Map