Assay Details
| Gene Symbol: Kcnmb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509082.2 | XP_003509130.1 | 2 - 3 | 235 | 81 | |
| XM_007649684.1 | XP_007647874.1 | 1 - 2 | 252 | 81 | ||
| XM_007649685.1 | XP_007647875.1 | 2 - 3 | 235 | 81 | ||
More Information
Set Membership: |
|

Kcnmb3
Genomic Map