Assay Details
| Gene Symbol: LOC100760681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649117.1 | XP_007647307.1 | 1 | 1022 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100760681| Gene Symbol: LOC100760681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649117.1 | XP_007647307.1 | 1 | 1022 | 111 | |
Set Membership: |
|