Assay Details
| Gene Symbol: Bnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500822.2 | XP_003500870.1 | 2 - 3 | 259 | 88 | |
| XM_007641563.1 | XP_007639753.1 | 2 - 3 | 243 | 88 | ||
| GenBank mRNA | JI870502.1 | - | 2 - 3 | 202 | 88 | |
| JP046517.1 | - | 2 - 3 | 202 | 88 | ||
More Information
Set Membership: |
|

Bnip1
Genomic Map