Assay Details
| Gene Symbol: Cd79b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501864.2 | XP_003501912.2 | 5 - 6 | 588 | 61 | |
| XM_007642522.1 | XP_007640712.1 | 4 - 5 | 279 | 61 | ||
More Information
Set Membership: |
|

Cd79b
Genomic Map