Assay Details
| Gene Symbol: Tprkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510022.2 | XP_003510070.1 | 1 - 2 | 235 | 73 | |
| GenBank mRNA | JI875734.1 | - | 1 - 2 | 89 | 73 | |
| JP038208.1 | - | 1 - 2 | 234 | 73 | ||
More Information
Set Membership: |
|

Tprkb
Genomic Map