Assay Details
| Gene Symbol: LOC100765263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648044.1 | XP_007646234.1 | 6 - 7 | 682 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100765263| Gene Symbol: LOC100765263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648044.1 | XP_007646234.1 | 6 - 7 | 682 | 79 | |
Set Membership: |
|