Assay Details
| Gene Symbol: LOC100752871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643276.1 | XP_007641466.1 | 3 - 4 | 1709 | 124 | |
| XM_007643281.1 | XP_007641471.1 | 3 - 4 | 1727 | 124 | ||
| XM_007643285.1 | XP_007641475.1 | 3 - 4 | 1926 | 124 | ||
| GenBank mRNA | JI883555.1 | - | 2 - 3 | 299 | 124 | |
More Information
Set Membership: |
|

LOC100752871
Genomic Map