Assay Details
| Gene Symbol: Haus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641745.1 | XP_007639935.1 | 4 - 5 | 630 | 65 | |
| GenBank mRNA | JI891254.1 | - | 4 - 5 | 630 | 65 | |
More Information
Set Membership: |
|

Haus1
Genomic Map