Assay Details
| Gene Symbol: LOC100771856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498154.2 | XP_003498202.1 | 4 - 5 | 616 | 65 | |
| XM_007622997.1 | XP_007621187.1 | 5 - 6 | 645 | 65 | ||
| XM_007623030.1 | XP_007621220.1 | 4 - 5 | 613 | 65 | ||
More Information
Set Membership: |
|

LOC100771856
Genomic Map