Assay Details
| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503924.2 | XP_003503972.2 | 16 - 17 | 2320 | 58 | |
| GenBank mRNA | JP045914.1 | - | 5 - 6 | 557 | 58 | |
More Information
Set Membership: |
|

Fam188b
Genomic Map