Assay Details
| Gene Symbol: Setd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509576.2 | XP_003509624.1 | 6 - 7 | 1160 | 63 | |
| XM_007650193.1 | XP_007648383.1 | 6 - 7 | 1099 | 63 | ||
| GenBank mRNA | JP057539.1 | - | 4 - 5 | 562 | 63 | |
More Information
Set Membership: |
|

Setd6
Genomic Map