Assay Details
| Gene Symbol: Ttc36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510512.2 | XP_003510560.1 | 1 - 2 | 561 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc36| Gene Symbol: Ttc36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510512.2 | XP_003510560.1 | 1 - 2 | 561 | 66 | |
Set Membership: |
|