Assay Details
| Gene Symbol: Nt5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495629.2 | XP_003495677.1 | 5 - 6 | 1107 | 65 | |
| XM_007642785.1 | XP_007640975.1 | 5 - 6 | 1107 | 65 | ||
More Information
Set Membership: |
|

Nt5e
Genomic Map