Assay Details
| Gene Symbol: Oma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495733.2 | XP_003495781.2 | 1443 | 67 | ||
| GenBank mRNA | JP054375.1 | - | 1443 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oma1| Gene Symbol: Oma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495733.2 | XP_003495781.2 | 1443 | 67 | ||
| GenBank mRNA | JP054375.1 | - | 1443 | 67 | ||
Set Membership: |
|