Assay Details
| Gene Symbol: Wipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510481.2 | XP_003510529.1 | 4 - 5 | 430 | 62 | |
| XM_007651112.1 | XP_007649302.1 | 4 - 5 | 432 | 62 | ||
| GenBank mRNA | JP055089.1 | - | 2 - 3 | 252 | 62 | |
More Information
Set Membership: |
|

Wipf2
Genomic Map