Assay Details
Gene Symbol: Rexo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509320.2 | XP_003509368.1 | 1 - 2 | 206 | 127 | |
XM_007649893.1 | XP_007648083.1 | 1 - 2 | 210 | 127 | ||
GenBank mRNA | JI891967.1 | - | 1 - 2 | 168 | 127 | |
JP057804.1 | - | 1 - 2 | 168 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
