Assay Details
| Gene Symbol: Ogfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643787.1 | XP_007641977.1 | 1 - 2 | 370 | 92 | |
| GenBank mRNA | JI896721.1 | - | 1 - 2 | 182 | 92 | |
More Information
Set Membership: |
|

Ogfr
Genomic Map