Assay Details
| Gene Symbol: Lyz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497141.2 | XP_003497189.1 | 2 - 3 | 304 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyz| Gene Symbol: Lyz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497141.2 | XP_003497189.1 | 2 - 3 | 304 | 105 | |
Set Membership: |
|