Assay Details
| Gene Symbol: Trub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513289.2 | XP_003513337.1 | 1 - 2 | 238 | 88 | |
| GenBank mRNA | JI887495.1 | - | 1 - 2 | 228 | 88 | |
| JP060278.1 | - | 1 - 2 | 228 | 88 | ||
More Information
Set Membership: |
|

Trub1
Genomic Map