Assay Details
| Gene Symbol: Krt81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654554.1 | XP_007652744.1 | 8 - 9 | 1284 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt81| Gene Symbol: Krt81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654554.1 | XP_007652744.1 | 8 - 9 | 1284 | 108 | |
Set Membership: |
|