Assay Details
| Gene Symbol: LOC103162367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007655117.1 | XP_007653307.1 | 4 | 1122 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103162367| Gene Symbol: LOC103162367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007655117.1 | XP_007653307.1 | 4 | 1122 | 90 | |
Set Membership: |
|