Assay Details
| Gene Symbol: Siah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508309.2 | XP_003508357.2 | 2 | 699 | 85 | |
| XM_007648939.1 | XP_007647129.1 | 2 | 588 | 85 | ||
| XM_007648940.1 | XP_007647130.1 | 2 | 519 | 85 | ||
More Information
Set Membership: |
|

Siah1
Genomic Map