Assay Details
| Gene Symbol: Abcf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501290.2 | XP_003501338.1 | 5 | 722 | 69 | |
| GenBank mRNA | JI876528.1 | - | 4 | 724 | 69 | |
| JP037949.1 | - | 4 | 724 | 69 | ||
More Information
Set Membership: |
|

Abcf2
Genomic Map