Assay Details
| Gene Symbol: Pigc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497658.2 | XP_003497706.1 | 2 | 1006 | 106 | |
| XM_007615363.1 | XP_007613553.1 | 2 | 1018 | 106 | ||
| XM_007615372.1 | XP_007613562.1 | 2 | 1011 | 106 | ||
| XM_007615379.1 | XP_007613569.1 | 2 | 968 | 106 | ||
More Information
Set Membership: |
|

Pigc
Genomic Map