Assay Details
| Gene Symbol: Birc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007633873.1 | XP_007632063.1 | 3 | 823 | 88 | |
| GenBank mRNA | JI877753.1 | - | 3 | 863 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Birc5| Gene Symbol: Birc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007633873.1 | XP_007632063.1 | 3 | 823 | 88 | |
| GenBank mRNA | JI877753.1 | - | 3 | 863 | 88 | |
Set Membership: |
|