Assay Details
| Gene Symbol: U2af1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645002.1 | XP_007643192.1 | 1 | 818 | 74 | |
| GenBank mRNA | JP048103.1 | - | 5 | 552 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

U2af1| Gene Symbol: U2af1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645002.1 | XP_007643192.1 | 1 | 818 | 74 | |
| GenBank mRNA | JP048103.1 | - | 5 | 552 | 74 | |
Set Membership: |
|