Assay Details
| Gene Symbol: Ptgfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496432.1 | XP_003496480.1 | 1 | 422 | 82 | |
| XM_007650087.1 | XP_007648277.1 | 1 | 422 | 82 | ||
More Information
Set Membership: |
|

Ptgfr
Genomic Map