Assay Details
| Gene Symbol: Pigp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648599.1 | XP_007646789.1 | 1 | 319 | 104 | |
| GenBank mRNA | JI874309.1 | - | 1 | 64 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pigp| Gene Symbol: Pigp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648599.1 | XP_007646789.1 | 1 | 319 | 104 | |
| GenBank mRNA | JI874309.1 | - | 1 | 64 | 104 | |
Set Membership: |
|