Assay Details
| Gene Symbol: LOC100775029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607640.1 | XP_007605830.1 | 4 | 1922 | 133 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100775029| Gene Symbol: LOC100775029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607640.1 | XP_007605830.1 | 4 | 1922 | 133 | |
Set Membership: |
|